Learn More
Description
Specifications
Specifications
| Antigen | GR/NR3C1 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | C.15200010 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.5 ug/mL, ELISA 0.5 ug/mL, Immunohistochemistry 2.5 ug/mL, Immunocytochemistry/Immunofluorescence 2.5 ug/mL, Immunoprecipitation 5 ug/IP, Gel Supershift Assay 5 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/IP, Chromatin Immunoprecipitation Sequencing 5ug/ChIP |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | GCR, glucocorticoid receptor, GRGCCR, GRLNuclear receptor subfamily 3 group C member 1, nuclear receptor subfamily 3, group C, member 1, nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor) |
| Gene Symbols | NR3C1 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
