Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRAMD1A Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB289943 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289943 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289943 Supplier Novus Biologicals Supplier No. NBP232148APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GRAMD1A Polyclonal antibody specifically detects GRAMD1A in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen GRAMD1A
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias FLJ22411, FLJ90346, GRAM domain containing 1A, GRAM domain-containing protein 1A, KIAA1533
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 674 to 724 of human GRAM Domain Containing 1A using the numbering given in entry NP_065946.2
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57655
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.