Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRAMD1A Antibody, Novus Biologicals™
SDP

Catalog No. NBP232147 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232147 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP232147 Supplier Novus Biologicals Supplier No. NBP232147
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GRAMD1A Polyclonal specifically detects GRAMD1A in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen GRAMD1A
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2 - 10 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_065946.2
Gene Alias FLJ22411, FLJ90346, GRAM domain containing 1A, GRAM domain-containing protein 1A, KIAA1533
Gene Symbols GRAMD1A
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 300 to 350 of human GRAM Domain Containing 1A using the numbering given in entry NP_065946.2 (GeneID 57655).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57655
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.