Learn More
Description
Specifications
Specifications
| Antigen | GRAMD3 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 350 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | GRAM domain containing 3, GRAM domain-containing protein 3, HCV NS3-transactivated protein 2, hepatitis C virus nonstructural protein 3-transactivating protein 2, NS3TP2FLJ21313 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human GRAMD3 fragment. |
| Purification Method | Antigen Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
