Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRAMD4 Antibody (PCRP-GRAMD4-1A10), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB157852 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB157852 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB157852 Supplier Novus Biologicals Supplier No. NBP314282R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GRAMD4 Monoclonal antibody specifically detects GRAMD4 in Human samples. It is validated for Flow Cytometry, Immunofluorescence

Specifications

Antigen GRAMD4
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PCRP-GRAMD4-1A10
Conjugate DyLight 550
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias GRAM domain containing 4, GRAM domain-containing protein 4, KIAA0767
Host Species Mouse
Immunogen Recombinant full-length human GRAMD4 protein (Uniprot: Q6IC98)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23151
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.