Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granulocyte Marker Antibody (SPM250), HRP, Novus Biologicals™
SDP

Catalog No. NBP234750H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234750H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234750H Supplier Novus Biologicals Supplier No. NBP234750H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Granulocyte Marker Monoclonal specifically detects Granulocyte Marker in Human, Rhesus Macaque samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Granulocyte Marker
Applications ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone SPM250
Conjugate HRP
Dilution ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Host Species Mouse
Immunogen Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes an unidentified antigen in the cytoplasm of mature granulocytes. It shows no reactivity with any other cell type in human tissues. Markers of myeloid cells are useful in the identification of different levels of cellular differentiation. monoclonal antibody BM-2 reacts with early precursor and mature forms of human myeloid cells. This monoclonal antibody is useful for the detection of myeloid leukemias and granulocytic sarcomas. It can be used as a marker of granulocytes in normal tissues or inflammatory processes.
Target Species Human, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.