Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granulocytes Antibody (HIS48), Novus Biologicals™
SDP

Catalog No. NB10064933 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064933 1 mL
NB017668 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10064933 Supplier Novus Biologicals Supplier No. NB10064933
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Granulocytes Monoclonal specifically detects Granulocytes in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).

Specifications

Antigen Granulocytes
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HIS48
Conjugate Unconjugated
Dilution Flow Cytometry Neat, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:20
Gene Alias C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3OS, filgrastim, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin
Gene Symbols CSF3
Host Species Mouse
Immunogen PVG rat spleen cell suspension
Purification Method Tissue culture supernatant
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1440
Test Specificity Recognizes granulocytes and erythroid cells. Clone HIS48 has frequently been used to stain rat neutrophils in immunohistochemistry (3).
Target Species Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.