Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granulysin Antibody (B-L38), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB388048 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB388048 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB388048 Supplier Novus Biologicals Supplier No. NBP318104AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Granulysin Monoclonal antibody specifically detects Granulysin in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen Granulysin
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone B-L38
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias D2S69Elymphocyte-activation gene 2, granulysin, LAG2, LAG-2, Lymphokine LAG-2, NKG5519, Protein NKG5, TLA519T-cell activation protein 519, T-lymphocyte activation gene 519
Host Species Mouse
Immunogen Recombinant human Granulysin (NKG5 and LAG-2). Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10578
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.