Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granulysin Antibody (B-R32), PE, Novus Biologicals™
SDP

Catalog No. NB340618 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340618 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340618 Supplier Novus Biologicals Supplier No. NBP318105PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Granulysin Monoclonal antibody specifically detects Granulysin in Human samples. It is validated for ELISA

Specifications

Antigen Granulysin
Applications ELISA
Classification Monoclonal
Clone B-R32
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias D2S69Elymphocyte-activation gene 2, granulysin, LAG2, LAG-2, Lymphokine LAG-2, NKG5519, Protein NKG5, TLA519T-cell activation protein 519, T-lymphocyte activation gene 519
Host Species Mouse
Immunogen Recombinant human Granulysin (NKG5 and LAG-2). Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10578
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.