Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Granulysin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB31381100
This item is not returnable.
View return policy
Description
Granulysin Monoclonal antibody specifically detects Granulysin in Human samples. It is validated for ELISASpecifications
Granulysin | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
D2S69Elymphocyte-activation gene 2, GNLY, granulysin, LAG2, LAG-2, Lymphokine, Lymphokine LAG-2, NKG5, NKG5519, Protein NKG5, TLA519, TLA519T-cell activation protein 519, T-lymphocyte activation gene 519 | |
Mouse myeloma cell line NS0-derived recombinant human Granulysin, Arg23-Leu145, Accession # P22749 | |
100 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2a |
ELISA | |
786314 | |
ELISA | |
P22749 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10578 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction