Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granzyme A Antibody (356412), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB363817 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363817 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363817 Supplier Novus Biologicals Supplier No. FAB29051J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Granzyme A Monoclonal specifically detects Granzyme A in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunoprecipitation.

Specifications

Antigen Granzyme A
Applications CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 356412
Conjugate Janelia Fluor 646
Dilution Immunoprecipitation, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CTL tryptase, CTLA3Granzyme A (Cytotoxic T-lymphocyte-associated serine esterase-3; Hanukah factorserine protease), Cytotoxic T-lymphocyte proteinase 1, EC 3.4.21, fragmentin-1, granzyme A, granzyme A (granzyme 1, cytotoxic T-lymphocyte-associated serine esterase 3), Granzyme-1, H factor, Hanukkah factor, HF, HFSPEC 3.4.21.78
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Granzyme A, Cys26-Val262, Accession # P12544
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Cancer, Immunology, Innate Immunity, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 3001
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.