Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granzyme B Antibody (2), Alexa Fluor™ 350
SDP

Catalog No. NB107637 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB107637 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB107637 Supplier Novus Biologicals Supplier No. NBP289439AF350

Rabbit Monoclonal Antibody

Granzyme B Monoclonal specifically detects Granzyme B in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Granzyme B
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GranzymeB, GZMB, HLP, SECT
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Granzyme B (NP_004122.1; Met 1-Tyr 247) .
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 3002
Test Specificity Has cross-reactivity in ELISA with: Human Granzyme H
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.