Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granzyme B Antibody (CLB-GB11), Novus Biologicals™
SDP

Catalog No. NBP150071 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP150071 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP150071 Supplier Novus Biologicals Supplier No. NBP150071
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Granzyme B Monoclonal specifically detects Granzyme B in Human, Primate samples. It is validated for Flow Cytometry, Flow (Intracellular), CyTOF-ready.

Specifications

Antigen Granzyme B
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone CLB-GB11
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1-4 ug/ml, Flow (Intracellular) 1-4 ug/ml, CyTOF-ready
Gene Alias C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GranzymeB, GZMB, HLP, SECT
Gene Symbols GZMB
Host Species Mouse
Immunogen Human NK cell line YT-INDY-derived granzyme B
Molecular Weight of Antigen 31 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3002
Test Specificity The mouse monoclonal antibody CLB-GB11 recognizes granzyme B, a 31 kDa serine protease expressed in activated Tc cells and NK cells.
Target Species Human, Primate
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.