Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granzyme B Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB283332 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB283332 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB283332 Supplier Novus Biologicals Supplier No. NBP259678MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Granzyme B Polyclonal antibody specifically detects Granzyme B in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Granzyme B
Applications Flow Cytometry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GranzymeB, GZMB, HLP, SECT
Host Species Rabbit
Immunogen Partial recombinant protein made to the human Granzyme B protein (amino acids 19-247) [UniProt P10144]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 3002
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.