Learn More
Granzyme D Antibody (RM0246-8G14), Alexa Fluor™ 350, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Granzyme D |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | RM0246-8G14 |
| Conjugate | Alexa Fluor 350 |
| Dilution | Western Blot |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | CCP5, Ctla5, Ctla-5, granzyme D |
| Gene Symbols | Gzmd |
| Host Species | Rat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.