Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granzyme K Antibody (2471A), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB305827 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305827 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305827 Supplier Novus Biologicals Supplier No. FAB10216L
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Granzyme K Monoclonal specifically detects Granzyme K in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Western Blot.

Specifications

Antigen Granzyme K
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2471A
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, Fragmentin-3, granzyme 3, granzyme K, granzyme K (granzyme 3, tryptase II), granzyme K (serine protease, granzyme 3, tryptase II), Granzyme-3, NK-Tryp-2, NK-tryptase-2, PRSS, TRYP2granzyme-3, tryptase II
Host Species Rabbit
Immunogen Synthetic peptide containing human Granzyme K, Accession # P49863
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3003
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.