Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Granzyme K Monoclonal Antibody (C8), Invitrogen™

Mouse Monoclonal Antibody
$195.80 - $429.50
Specifications
Antigen | Granzyme K |
---|---|
Clone | C8 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEMA551115
![]() |
Invitrogen™
MA551115 |
200 μL |
Each for $429.50
|
|
|||||
FEMA551116
|
Invitrogen™
MA551116 |
20 μL |
Each for $195.80
|
|
|||||
Description
Granzyme K (GrK) belongs to a family of trypsin-like serine proteases localised in the cytotoxic granules of activated T cells and NK cells. It encodes a 28 kDa serine protease whose gene is located on chromosome 5q11-12 close to the granzyme A-encoding gene. Like granzyme A, it has a trypsin-like specifity cleaving at the basic residues arginine and lysine. Granzyme K triggers rapid cell death independently of caspase activation with single-stranded DNA nicks and is primarily expressed in thymus, lung, spleen and peripheral blood leukocytes.Specifications
Granzyme K | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
Fragmentin-3; granzyme 3; granzyme K; granzyme K (granzyme 3; tryptase II); granzyme K (serine protease, granzyme 3; tryptase II); granzyme-3; GrK; GZMK; NK-Tryp-2; NK-tryptase-2; RNK-Tryp-2; TRYP2; tryptase II | |
GZMK | |
IgG2b κ | |
Protein A/G | |
GZMK |
C8 | |
Western Blot | |
Unconjugated | |
Mouse | |
Mouse, Rat | |
O35205, P49864 | |
14945, 29165 | |
Recombinant Mouse GZMK protein, Gln44-Val227 (Accession #O35205). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title