Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRAP2 Antibody (OTI1G2), HRP, Novus Biologicals™
SDP

Catalog No. NBP271457H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271457H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271457H Supplier Novus Biologicals Supplier No. NBP271457H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GRAP2 Monoclonal specifically detects GRAP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen GRAP2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1G2
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias Adapter protein GRID, GADSSH3-SH2-SH3 adapter Mona, GRB2L, GRB-2-like protein, GRB2-related adapter protein 2, GRB2-related adaptor protein 2, GRB2-related protein with insert domain, GRBLGGRAP-2, GRBX, grf-40, Grf40, Grf40 adapter protein, GRIDGRPL, Growth factor receptor-binding protein, growth factor receptor-bound protein 2-related adaptor protein 2, Hematopoietic cell-associated adapter protein GrpL, hematopoietic cell-associated adaptor protein GRPL, Mona, P38, Protein GADS, SH3-SH2-SH3 adaptor molecule
Gene Symbols GRAP2
Host Species Mouse
Immunogen Full length recombinant protein of human GRAP2 (NP_004801) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 9402.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.