Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRASP1 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001432 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001432 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001432 Supplier Novus Biologicals Supplier No. NB1001432
Add to Cart
Add to Cart

Goat Polyclonal Antibody

GRASP1 Polyclonal specifically detects GRASP1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot.

Specifications

Antigen GRASP1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Immunohistochemistry-Paraffin 3.75 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_064522
Gene Alias DKFZp434P0630, GRASP1, GRASP-1MGC126595, GRIP1 associated protein 1, GRIP1-associated protein 1, KIAA1167MGC126593, MPMGp800B12492Q3
Gene Symbols GRIPAP1
Host Species Goat
Immunogen Peptide with sequence LREQTTGLAAELQ-C corresponding to N-Terminus according to NP_064522.3, NP_997555.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 56850
Test Specificity This antibody is expected to recognize both reported isoforms (as represented by NP_064522.3; NP_997555.1)
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.