Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRASP1 Antibody (OTI1E3), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB272064APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272064APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272064APC Supplier Novus Biologicals Supplier No. NBP272064APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GRASP1 Monoclonal specifically detects GRASP1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen GRASP1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1E3
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias DKFZp434P0630, GRASP1, GRASP-1MGC126595, GRIP1 associated protein 1, GRIP1-associated protein 1, KIAA1167MGC126593, MPMGp800B12492Q3
Gene Symbols GRIPAP1
Host Species Mouse
Immunogen Full length recombinant protein of human GRASP1 (NP_064522) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 56850.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.