Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRASP65 Antibody (OTI5G8), PE, Novus Biologicals™
SDP

Catalog No. NBP272144PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272144PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272144PE Supplier Novus Biologicals Supplier No. NBP272144PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GRASP65 Monoclonal specifically detects GRASP65 in Human, Rat, Porcine, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.

Specifications

Antigen GRASP65
Applications Flow Cytometry
Classification Monoclonal
Clone OTI5G8
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Golgi peripheral membrane protein p65, Golgi phosphoprotein 5MGC118897, golgi reassembly stacking protein 1, 65kDa, Golgi reassembly-stacking protein 1, Golgi reassembly-stacking protein of 65 kDa, GRASP65MGC118894, P6565 kDa
Gene Symbols GORASP1
Host Species Mouse
Immunogen Full length recombinant protein of human GRASP65 (NP_114105) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 64689.0
Target Species Human, Rat, Pig, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.