Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRASP65/GORASP1 Mouse anti-Human, Mouse, Rat, Clone: 1B2C10, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | GRASP65/GORASP1 |
---|---|
Clone | 1B2C10 |
Concentration | 0.8 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7069
![]() ![]() |
Proteintech Group Inc
666511IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Grasp65 is a structural protein of the golgi apparatus. It has been implicated in the stacking of golgi cisternae. Grasp65 is shown to have the properties to bind surfaces together in a mitotically regulated matter.Specifications
GRASP65/GORASP1 | |
0.8 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FLJ23443, Golgi phosphoprotein 5, GOLPH5, GORASP1, GRASP65, P65 | |
GORASP1 | |
IgG1 | |
Protein G | |
Antibody |
1B2C10 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
O35254, Q91X51, Q9BQQ3 | |
56082, 64689, 74498 | |
GRASP65,GORASP1 Fusion Protein Ag1054 | |
Primary | |
-20°C | |
GORASP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title