Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501726154 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6154 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6154 Supplier Proteintech Group Inc Supplier No. 102542AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GRB 2 protein contains one SH2 domain flanked on both sides by an SH3 domain. In response to EGF, GRB 2 binds, through its SH3 domains, to the guanine nucleotide exchange factor, Sos. The GRB 2 - Sos complex then binds the EGF receptor. These interactions play a critical role in the activation of the p21 ras signaling pathway.

Specifications

Antigen GRB2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene GRB2
Gene Accession No. P62993, P62994, Q60631
Gene Alias Adapter protein GRB2, ASH, EGFRBP GRB2, GRB2, Grb3 3, MST084, MSTP084, Protein Ash, SH2/SH3 adapter GRB2
Gene Symbols GRB2
Host Species Rabbit
Immunogen GRB2 Fusion Protein Ag0395
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14784, 2885, 81504
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.