Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Green Caps For Lysing Matrix Tubes

Catalog No. 115067005 Shop All MP Biomedicals, Inc Products
Click to view available options
Color:
Green

Catalog No. 11-506-7005

Supplier: MP Biomedicals™ 115067005

Only null left
Add to Cart
Add to Cart

Colored screw cap with o-ring fits empty 2 mL microfuge tubes. Green Caps for Lysing Matrix Tubes are used in conjunction for isolation of total RNA from plants and animals.

  • Fit empty 2 mL FastPrep tubes
  • For use in FastPrep systems
  • Available separately for those who wish to fill there own lysing matrix tubes.
  • Applications include sample preparation, sample lysis, nucleic acid isolation and purification, protein isolation, molecular biology, gene expression, microbiological analysis, gene function, gene regulation
TRUSTED_SUSTAINABILITY

Specifications

Includes O-ring
Product Type Screw Cap
Color Green
Quantity 1000 Pk.
Capacity (Metric) 2 mL
For Use With (Equipment) FastRNA™ Pro Green Lysing Matrix D Tubes
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.