Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gremlin 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
Supplier: Novus Biologicals NB10056883
Description
Gremlin 1 Polyclonal specifically detects Gremlin 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Gremlin 1 | |
Polyclonal | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
Cell proliferation-inducing gene 2 protein, CKTSF1B1gremlin 1, cysteine knot superfamily, homolog, Cysteine knot superfamily 1, BMP antagonist 1gremlin 1, cysteine knot superfamily, homolog (Xenopus laevis), DAN domain family member 2, DAND2GREMLIN, Down-regulated in Mos-transformed cells protein, DRMMGC126660, gremlin, gremlin 1, gremlin 1-like protein, gremlin-1, IHG-2, Increased in high glucose protein 2, increased in high glucose-2, proliferation-inducing gene 2 | |
Rabbit | |
Protein G purified | |
RUO | |
Primary | |
Human. Predicted cross-reactivity based on sequence identity: Mouse (94%) and Rat (94%). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
1.0 mg/mL | |
Immunohistochemistry 1:20-1:1000, Immunohistochemistry-Paraffin 2.5-10 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (Namkoong et al (2006)) | |
O60565 | |
GREM1 | |
A synthetic peptide containing amino acids QPGSRNRGRGQGRGTA (52-67) of human CKTSF1B1 (NM_013372). | |
0.1 mg | |
Neuroscience | |
26585 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction