Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gremlin 1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 5 publications

$206.50 - $499.50

Specifications

Antigen Gremlin 1
Concentration 1.0 mg/mL
Dilution Immunohistochemistry 1:20-1:1000, Immunohistochemistry-Paraffin 2.5-10 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (Namkoong et al (2006))
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056883T
SDP
View Documents
Novus Biologicals
NB10056883SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056883
SDP
View Documents
Novus Biologicals
NB10056883
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Gremlin 1 Polyclonal specifically detects Gremlin 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Gremlin 1
Immunohistochemistry 1:20-1:1000, Immunohistochemistry-Paraffin 2.5-10 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (Namkoong et al (2006))
Polyclonal
Purified
RUO
Human
O60565
26585
A synthetic peptide containing amino acids QPGSRNRGRGQGRGTA (52-67) of human CKTSF1B1 (NM_013372).
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1.0 mg/mL
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
Rabbit
Neuroscience
PBS with 0.05% Sodium Azide
Cell proliferation-inducing gene 2 protein, CKTSF1B1gremlin 1, cysteine knot superfamily, homolog, Cysteine knot superfamily 1, BMP antagonist 1gremlin 1, cysteine knot superfamily, homolog (Xenopus laevis), DAN domain family member 2, DAND2GREMLIN, Down-regulated in Mos-transformed cells protein, DRMMGC126660, gremlin, gremlin 1, gremlin 1-like protein, gremlin-1, IHG-2, Increased in high glucose protein 2, increased in high glucose-2, proliferation-inducing gene 2
GREM1
IgG
Protein G purified
Human. Predicted cross-reactivity based on sequence identity: Mouse (94%) and Rat (94%).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.