Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRG (Groucho homolog) Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB093615 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093615 20 μL
NB093614 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB093615 Supplier Novus Biologicals Supplier No. NBP30340520UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GRG (Groucho homolog) Polyclonal specifically detects GRG (Groucho homolog) in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GRG (Groucho homolog)
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias AES-1, AES-2, Amino enhancer of split, amino-terminal enhancer of split, ESP1, Gp130-associated protein GAM, GRG, GRG5, Protein ESP1, Protein GRG, TLE5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-264 of human GRG (Groucho homolog) (NP_945320.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Apoptosis, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 166
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.