Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRHL1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB350333 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350333 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350333 Supplier Novus Biologicals Supplier No. NBP321433MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GRHL1 Polyclonal antibody specifically detects GRHL1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen GRHL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias grainyhead-like 1 (Drosophila), grainyhead-like protein 1 homolog, LBP protein 32, MGRLBP-32, NH32, TFCP2L2, Transcription factor CP2-like 2LBP32Mammalian grainyhead, Transcription factor LBP-32
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human GRHL1 fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29841
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.