Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRHPR Antibody (OTI2H4), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N271430A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271430A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271430A405 Supplier Novus Biologicals Supplier No. NBP271430AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GRHPR Monoclonal specifically detects GRHPR in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen GRHPR
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI2H4
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 1.1.1.81, GLXREC 1.1.1.79, glycerate-2-dehydrogenase, GLYD, glyoxylate reductase/hydroxypyruvate reductase, PH2
Gene Symbols GRHPR
Host Species Mouse
Immunogen Full length recombinant protein of human GRHPR (NP_036335) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 9380.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.