Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRID2 Antibody, Novus Biologicals™
SDP

Catalog No. NB018619 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018619 0.025 mL
NBP146489 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018619 Supplier Novus Biologicals Supplier No. NBP1464890.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GRID2 Polyclonal specifically detects GRID2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen GRID2
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation Whole antisera with No Preservative
Gene Alias B230104L07Rik, cpr, creeper, gluR delta-2 subunit, GluRdelta2, glutamate receptor delta-2 subunit, glutamate receptor, ionotropic, delta 2, ho, hotfoot, Lc, lurcher, minisatellite 10ac detected by probe MMS10, MMS10-AC, Ms10ac, nmf408, tpr
Gene Symbols GRID2
Host Species Rabbit
Immunogen A synthetic peptide from c-terminal region of human GRID2 (Glutamate receptor delta-2 subunit) conjugated to an immunogenic carrier protein was used as the antigen.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 2895
Test Specificity Specific for GRID2.
Reconstitution Reconstitute with 0.1 ml sterilized water to desired concentration.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.