Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRID2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
| Antigen | GRID2 |
|---|---|
| Concentration | LYOPH |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
GRID2 Polyclonal specifically detects GRID2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| GRID2 | |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| Neuronal Cell Markers, Neurotransmission | |
| B230104L07Rik, cpr, creeper, gluR delta-2 subunit, GluRdelta2, glutamate receptor delta-2 subunit, glutamate receptor, ionotropic, delta 2, ho, hotfoot, Lc, lurcher, minisatellite 10ac detected by probe MMS10, MMS10-AC, Ms10ac, nmf408, tpr | |
| GRID2 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| LYOPH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat | |
| 2895 | |
| IgG | |
| Unpurified | |
| Specific for GRID2. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title