Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRIK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7205245 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8863 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8863 Supplier Proteintech Group Inc Supplier No. 135971AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Glutamate receptors are the predomit excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. Grik2, also known as glutamate receptor 6, belongs to the kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels. Recent reports have suggested that defects in the Grik2 protein may be associated with autosomal recessive mental retardation and possibly other neurological disorders such as schizophrenia. Numerous isoforms of Grik2 are known to exist and may be subject to RNA editing within the second transmembrane domain, which is thought to alter the properties of ion flow. This Grik2 antibody may exhibit some cross-reactivity to Grik3.

Specifications

Antigen GRIK2
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GRIK2
Gene Accession No. P39087, P42260, Q13002
Gene Alias EAA4, GLR6, GluK2, GluR 6, GLUR6, Glutamate receptor 6, GRIK2, MRT6
Gene Symbols GRIK2
Host Species Rabbit
Immunogen GRIK2 Fusion Protein Ag4516
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14806, 2898, 54257
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.