Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRIM19 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB104272 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB104272 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB104272 Supplier Novus Biologicals Supplier No. NBP298488AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GRIM19 Polyclonal specifically detects GRIM19 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GRIM19
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias B16.6, CDA016, Cell death regulatory protein GRIM-19, cell death-regulatory protein GRIM19, CI-B16.6, complex I B16.6 subunit, Complex I-B16.6, FLJ58045, FLJ59191, Gene associated with retinoic and IFN-induced mortality 19 protein, Gene associated with retinoic and interferon-induced mortality 19 protein, GRIM-19CGI-39, GRIM19NADH-ubiquinone oxidoreductase B16.6 subunit, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human GRIM19 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 51079
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.