Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPEX™ SamplePrep Grinding Vial Accessory Packages

Catalog No. p-4422562
Click to view available options
Size:
Small
Large

Contains everything necessary to run Small/Mid-size/Large grinding vials in the LargeFreezer/Mill™

  • Closed grinding vial prevents cross-contamination.
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles.
  • Compatible with grinding vial sets 61043-09 and 61043-11.
  • Compatible with Freezer/Mill™ cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.

Specifications

Product Type Grinding Set
For Use With (Equipment) Freezer/Mill Cryogenic Grinders
Includes 1 Vial Extractor/Opener (04576-38) and Vial Rack (04576-39)
Size Large
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.