Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRIP1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB359535 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359535 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359535 Supplier Novus Biologicals Supplier No. NB1202842MFV450

Rabbit Polyclonal Antibody

GRIP1 Polyclonal antibody specifically detects GRIP1 in Human, Hamster samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen GRIP1
Applications Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias glutamate receptor interacting protein 1, glutamate receptor-interacting protein 1, GRIP, GRIP-1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues P(206) L P D S E E E G H D S(217) C of human GRIP1.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuronal Cell Markers, Neurotransmission, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 23426
Target Species Human, Hamster
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.