Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548489
Change view
Click to view available options
Quantity:
400 μL
Catalog No. Quantity
PA548489 400 μL
1 options

Catalog No. PA548489

Supplier: Invitrogen™ PA548489

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GRIP2 Polyclonal Antibody for Western Blot, IHC (P)

May play a role as a localized scaffold for the assembly of a multiprotein signaling complex and as mediator of the trafficking of its binding partners at specific subcellular location in neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRIP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GRIP2
Gene Accession No. Q9C0E4
Gene Alias glutamate receptor interacting protein 2; glutamate receptor-interacting protein 2; GRIP2; GRIP-2; KIAA1719
Gene Symbols GRIP2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 620-649 amino acids from the Central region of human GRIP2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80852
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.