Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRK1 Antibody (D11), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB359537 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359537 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359537 Supplier Novus Biologicals Supplier No. NB1202776MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GRK1 Monoclonal antibody specifically detects GRK1 in Human, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)

Specifications

Antigen GRK1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone D11
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias G protein-coupled receptor kinase 1rhodopsin kinase, GPRK1, RHOKEC 2.7.11, RKEC 2.7.11.14
Host Species Mouse
Immunogen Full length human GRK1.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6011
Target Species Human, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.