Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRK1 Antibody (G8), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB1202775Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1202775Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1202775Z Supplier Novus Biologicals Supplier No. NB1202775AF700

Mouse Monoclonal Antibody

GRK1 Monoclonal specifically detects GRK1 in Human, Mouse, Rat, Bovine, Chicken samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GRK1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone G8
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Gene Alias G protein-coupled receptor kinase 1rhodopsin kinase, GPRK1, RHOKEC 2.7.11, RKEC 2.7.11.14
Gene Symbols GRK1
Host Species Mouse
Immunogen Full length human GRK1.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6011
Test Specificity Detects G-protein-associated rhodopsin kinase 1a (GRK1a).
Target Species Human, Mouse, Rat, Bovine, Chicken
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.