Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRK2 Antibody, Novus Biologicals™
SDP

Catalog No. NB1002831 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002831 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002831 Supplier Novus Biologicals Supplier No. NB1002831
Add to Cart
Add to Cart

Goat Polyclonal Antibody

GRK2 Polyclonal specifically detects GRK2 in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen GRK2
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 2 ug/mL, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001610.1
Gene Alias adrenergic, beta, receptor kinase 1, BARK, BARK1beta-ARK-1, Beta-ARK-1, BETA-ARK1, EC 2.7.11, EC 2.7.11.15, FLJ16718, G-protein coupled receptor kinase 2, GRK2beta-adrenergic receptor kinase 1
Gene Symbols ADRBK1
Host Species Goat
Immunogen Peptide with sequence C-RDAYREAQQLVQR corresponding to internal region according to NP_001610.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Mitochondrial Markers, Neuroscience, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 156
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog, Pig and Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.