Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRK4 Antibody (OTI4F5), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP270845V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270845V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270845V Supplier Novus Biologicals Supplier No. NBP270845V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GRK4 Monoclonal specifically detects GRK4 in Human samples. It is validated for Western Blot.

Specifications

Antigen GRK4
Applications Western Blot
Classification Monoclonal
Clone OTI4F5
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.7.11, EC 2.7.11.16, G protein-coupled receptor kinase 2-like (Drosophila), G protein-coupled receptor kinase 4, G protein-coupled receptor kinase GRK4, GPRK2LIT11, GPRK4GRK4a, G-protein coupled receptor kinase 4, ITI1
Gene Symbols GRK4
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-245 of human GRK4 (NP_892027) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2868.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.