Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GroEL Antibody, Novus Biologicals™
SDP

Catalog No. NB036602 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036602 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036602 Supplier Novus Biologicals Supplier No. NBP289011
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GroEL Polyclonal specifically detects GroEL in E. coli samples. It is validated for Western Blot.

Specifications

Antigen GroEL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS containing 50% glycerol
Gene Alias Cpn60 chaperonin GroEL, large subunit of GroESL, ECK4137, groEL, JW4103, mopA
Host Species Rabbit
Immunogen E. coli GroEL.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 948665
Target Species E. coli
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.