Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Growth Hormone Antibody (101), FITC
SDP

Catalog No. NB072290 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB072290 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB072290 Supplier Novus Biologicals Supplier No. NBP290326F

Rabbit Monoclonal Antibody

Growth Hormone Monoclonal specifically detects Growth Hormone in Human samples. It is validated for ELISA.

Specifications

Antigen Growth Hormone
Applications ELISA
Classification Monoclonal
Clone 101
Conjugate FITC
Dilution ELISA
Formulation PBS
Gene Alias GHIGHD1B, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Growth Hormone (P01241; Phe27-Phe217).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 2688
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.