Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Growth Hormone Antibody (GH-45), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB500364549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500364549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500364549 Supplier Novus Biologicals Supplier No. NB500364JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Growth Hormone Monoclonal specifically detects Growth Hormone in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen GrowthHormone
Applications ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GH-45
Conjugate Janelia Fluor 549
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias GHIGHD1B, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N
Gene Symbols GH1
Host Species Mouse
Immunogen Human growth hormone
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 2688
Test Specificity The antibody GH-45 reacts with human growth hormone (hGH), a polypeptide hormone synthesized by acidophilic or somatotropic cells of the anterior pituitary gland. The GH-45 antibody reacts with affinity constant 3.8 x 1010 lmol; it does not bind human prolactin or any other pituitary hormones.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.