Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Growth Hormone R Antibody (014), DyLight 755
SDP

Catalog No. NB072767 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB072767 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB072767 Supplier Novus Biologicals Supplier No. NBP290375IR

Rabbit Monoclonal Antibody

Growth Hormone R Monoclonal specifically detects Growth Hormone R in Mouse samples. It is validated for ELISA.

Specifications

Antigen Growth Hormone R
Applications ELISA
Classification Monoclonal
Clone 14
Conjugate DyLight 755
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias GH receptor, GHBP, growth hormone binding protein, growth hormone receptor, serum binding protein, Somatotropin receptor
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Growth Hormone R (NP_034414.2; Met1-Gln273).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2690
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.