Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Growth Hormone R Antibody (10C11), Novus Biologicals™
SDP

Catalog No. NB297868 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297868 50 μL
NB297869 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297868 Supplier Novus Biologicals Supplier No. NBP32655450UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Growth Hormone R Monoclonal antibody specifically detects Growth Hormone R in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry

Specifications

Antigen Growth Hormone R
Applications ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 10C11
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias GH receptor, GHBP, growth hormone binding protein, growth hormone receptor, serum binding protein, Somatotropin receptor
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Growth Hormone R [UniProt P10912]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cytokine Research, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2690
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.