Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Growth Hormone R Antibody (10C11), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Growth Hormone R |
---|---|
Clone | 10C11 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
Applications | ELISA, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Description
Growth Hormone R Monoclonal antibody specifically detects Growth Hormone R in Human samples. It is validated for ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
Growth Hormone R | |
ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
GH receptor, GHBP, growth hormone binding protein, growth hormone receptor, serum binding protein, Somatotropin receptor | |
A synthesized peptide derived from Human Growth Hormone R [UniProt P10912] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
10C11 | |
ELISA, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Cytokine Research, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
2690 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title