Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRP75/HSPA9B/Mortalin Antibody (30A5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP1974480.025ML
Description
GRP75/HSPA9B/Mortalin Monoclonal specifically detects GRP75/HSPA9B/Mortalin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Drosophila, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GRP75/HSPA9B/Mortalin | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
HSPA9 | |
Native human Grp75 (Mortalin). | |
0.025 mL | |
Core ESC Like Genes, Stem Cell Markers | |
3313 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
30A5 | |
Western Blot 1:1000, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:10-1:500 | |
75 kDa glucose-regulated protein, GRP-75, GRP75mthsp75, Heat shock 70 kDa protein 9, heat shock 70kD protein 9B, heat shock 70kDa protein 9 (mortalin), heat shock 70kDa protein 9B (mortalin-2), HSPA9BMGC4500, Mortalin, mortalin, perinuclear, mortalin-2, MOT, mot-2, MOT2, MTHSP75, p66-mortalin, PBP74CSA, Peptide-binding protein 74, stress-70 protein, mitochondrial | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
Hamster, Pig, Human, Chicken, Guinea Pig, Rat, Sheep, Primate, Bovine, Rabbit, Xenopus species, Canine, Drosophila, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction