Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRP75/HSPA9B/Mortalin Antibody (30A5), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$304.50 - $828.50

Specifications

Antigen GRP75/HSPA9B/Mortalin
Clone 30A5
Dilution Western Blot 1:1000, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:10-1:500
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018765
SDP
View Documents
Novus Biologicals
NBP1974480.025ML
0.025 mL
Each for $304.50
Only null left
Add to Cart
 
NBP19744805
SDP
View Documents
Novus Biologicals
NBP1974480.05ML
0.05 mL
Each for $487.50
Only null left
Add to Cart
 
NBP19744802
SDP
View Documents
Novus Biologicals
NBP1974480.2ML
0.2 mL
Each for $828.50
Only null left
Add to Cart
 
Description

Description

GRP75/HSPA9B/Mortalin Monoclonal specifically detects GRP75/HSPA9B/Mortalin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Drosophila, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

GRP75/HSPA9B/Mortalin
Western Blot 1:1000, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:10-1:500
Monoclonal
Purified
RUO
Hamster, Pig, Human, Chicken, Guinea Pig, Rat, Sheep, Primate, Bovine, Rabbit, Xenopus species, Canine, Drosophila, Mouse
75 kDa glucose-regulated protein, GRP-75, GRP75mthsp75, Heat shock 70 kDa protein 9, heat shock 70kD protein 9B, heat shock 70kDa protein 9 (mortalin), heat shock 70kDa protein 9B (mortalin-2), HSPA9BMGC4500, Mortalin, mortalin, perinuclear, mortalin-2, MOT, mot-2, MOT2, MTHSP75, p66-mortalin, PBP74CSA, Peptide-binding protein 74, stress-70 protein, mitochondrial
HSPA9
IgG1
Ammonium sulfate precipitation
30A5
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Core ESC Like Genes, Stem Cell Markers
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
3313
Native human Grp75 (Mortalin).
Primary
Store at -20C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.