Learn More
GRP75/HSPA9B/Mortalin Antibody (OTI9F8), Alexa Fluor™ 700, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | GRP75/HSPA9B/Mortalin |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | OTI9F8 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | 75 kDa glucose-regulated protein, GRP-75, GRP75mthsp75, Heat shock 70 kDa protein 9, heat shock 70kD protein 9B, heat shock 70kDa protein 9 (mortalin), heat shock 70kDa protein 9B (mortalin-2), HSPA9BMGC4500, Mortalin, mortalin, perinuclear, mortalin-2, MOT, mot-2, MOT2, MTHSP75, p66-mortalin, PBP74CSA, Peptide-binding protein 74, stress-70 protein, mitochondrial |
| Gene Symbols | HSPA9 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.