Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRP78/HSPA5 Antibody (1H11-1H7), FITC, Novus Biologicals™
SDP

Catalog No. NBP212888F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212888F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212888F Supplier Novus Biologicals Supplier No. NBP212888F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GRP78/HSPA5 Monoclonal specifically detects GRP78/HSPA5 in Human, Mouse, Rat, Bovine, Fungi, Hamster, Monkey, Rabbit, Xenopus samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GRP78/HSPA5
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1H11-1H7
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias 78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2
Gene Symbols HSPA5
Host Species Mouse
Immunogen His-tagged human GRP78
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR)
Primary or Secondary Primary
Gene ID (Entrez) 3309
Test Specificity Detects a protein with a mass of approx. 78kDa corresponding to GRP78
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.